


Hydrogen chloride 2 N in diethyl ether, AcroSeal®, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Density:0.74 g/cm³
Flash Pt:–40 °C
EINECS:231-595-7
UN:2924
ADR:3,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:1.90 to 2.20 N
Specification Test Results
Related Information
Frequently Bought Together