Order Entry
Czech Republic
ContactUsLinkComponent
Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for AAS, Specpure®
Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for AAS, Specpure®
Catalog # 88096.AE
CAS Number:  
Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for AAS, Specpure®
Catalog # 88096.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00003397
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397

Certifications: Traceable to NIST