Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol)
Catalog # EHERC12570000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:370.49 g/mol
EINECS:204-082-0
CAS Number:115-32-2
Formula:C₁₄H₉Cl₅O