1,2-Dichloroethane-[D₄], Sigma-Aldrich®
: Merck
:17060-07-0
:
:C₂D₄Cl₂
:MFCD00037531
:102.93 g/mol
:3,II
:M-Clarity™ quality level = MQ200
:1184
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals