
Tris(2-ethylhexyl)phosphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-42-2
:
Formula:C₂₄H₅₁O₄P
MDL Number:MFCD00009491
:434.64 g/mol
Boiling Pt:216 °C (4 mmHg)
Density:0.93
Flash Pt:>110 °C (230 °F)
EINECS:201-116-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together