1,1'-Binaphthyl ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 604-53-5 Molecular Formula: C20H14 Formula Weight: 254.33 Melting Point: 144-146° MDL No.: MFCD00041740
Synonyms: 1,1'-Binaphthalene
- CAS Number:604-53-5
- Formula:C₂₀H₁₄
- MDL Number:MFCD00041740
- Molecular Weight:254.33 g/mol
- Boiling Pt:405 °C (1013 hPa)
- Melting Pt:144…146 °C
- Density:1.19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:210-070-6
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 604-53-5 Molecular Formula: C20H14 Formula Weight: 254.33 Melting Point: 144-146° MDL No.: MFCD00041740
Synonyms: 1,1'-Binaphthalene
- CAS Number:604-53-5
- Formula:C₂₀H₁₄
- MDL Number:MFCD00041740
- Molecular Weight:254.33 g/mol
- Boiling Pt:405 °C (1013 hPa)
- Melting Pt:144…146 °C
- Density:1.19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:210-070-6
Documents
Organic Building Blocks and Intermediates
Application Notes
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



