
2,4-Difluorobenzenesulphonamide 96%
Catalog # H55437.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:193.17 g/mol
Melting Pt:154…158 °C
:GHS07
Storage Temperature:Ambient
CAS Number:13656-60-5
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD01320743