Order Entry
Czech Republic
ContactUsLinkComponent
Silicon standard solution, 1,000 mg/L Si in water standard for AAS
Silicon standard solution, 1,000 mg/L Si in water standard for AAS
Catalog # REGEAASIH
Supplier:  REAGECON
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in water standard for AAS
Catalog # REGEAASIH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item