Potassium standard solution, 10,000 mg/L K in dilute nitric acid standard for ICP
Catalog # REGEPK4B2
Dodavatel: REAGECON
Číslo CAS:
Specifikace
- Concentration:10000mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:250 mL
- Element:Potassium
- Symbol:K