Order Entry
Czech Republic
ContactUsLinkComponent
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid standard for ICP
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid standard for ICP
Catalog # REGEPZN2A2
Supplier:  REAGECON
CAS Number:  
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid standard for ICP
Catalog # REGEPZN2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item