Order Entry
Czech Republic
ContactUsLinkComponent
 
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid standard for ICP
  REGEPNI2A2
 :  REAGECON
 :  
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid standard for ICP
  REGEPNI2A2
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Nickel
  • Symbol:
    Ni