Order Entry
Czech Republic
ContactUsLinkComponent
Cadmium standard solution, 1,000 mg/l Cd in 0.5 M nitric acid (from Cd(NO₃)₂) standard for AAS
Cadmium standard solution, 1,000 mg/l Cd in 0.5 M nitric acid (from Cd(NO₃)₂) standard for AAS
Catalog # SCARCA00410100
Supplier:  Scharlau
CAS Number:  
Cadmium standard solution, 1,000 mg/l Cd in 0.5 M nitric acid (from Cd(NO₃)₂) standard for AAS
Catalog # SCARCA00410100
Supplier:  Scharlau
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item