trans-4-Hexen-1-ol 97%
Catalog # H54459.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:100.16 g/mol
Boiling Pt:159…160 °C
Density:0.851
Flash Pt:61 °C (142 °F)
Storage Temperature:Ambient
EINECS:213-188-6
CAS Number:928-92-7
Formula:C₆H₁₂O
MDL Number:MFCD00009713