Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid (from Mg(NO₃)₂) standard for AAS
Catalog # SCARMA00110100
Supplier: Scharlau
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Magnesium
- Symbol:Mg