Order Entry
Czech Republic
ContactUsLinkComponent
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for ICP
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for ICP
Catalog # REGEPCD2A2
Supplier:  REAGECON
CAS Number:  
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for ICP
Catalog # REGEPCD2A2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item