ContactUsLinkComponent
CSB-Reaction Vessel
  ROBU18910
undefined
CSB-Reaction Vessel
  ROBU18910
  • Connection:
    NSH 29/32
  • Ext.Ø:
    40 mm
  • Description:
    CSB-reaction vessel
  • Pk:
    2
ROBU® CSB-reaction vessel with VitraPOR® filter is a high-quality, durable vessel designed for chemical reaction processes in laboratory and industrial applications. Made from resistant materials, it ensures precise control and reliable performance during reactions requiring stability and temperature resistance.
  • Durable construction
  • Resistant to high temperatures and chemicals
  • Ideal for chemical reaction processes
  • Precise control of reaction conditions
  • Easy integration into laboratory setups
  • Reusable and autoclavable
  • Ensures safe and efficient reaction management
ROBU CSB-reaction vessel with intergrated VitraPOR filter is specifically designed for use in chemical reaction processes, offering outstanding durability and chemical resistance. Whether used in laboratory experiments or industrial applications, it provides an efficient solution for controlling and managing chemical reactions under varying conditions. Made from high-quality borosilicate, it is resistant to high temperatures and chemicals, ensuring that it can withstand the rigors of demanding reaction environments. The vessel’s precise design allows for accurate control over reaction conditions, making it a reliable choice for researchers and industry professionals. It is reusable, autoclavable, and designed for easy integration into existing setups, offering long-term value and efficiency.
Standard ground joint: NSH 29/32
Filter Ø: 40 mm
 :
In compliance with ISO 4796-80 and ASTM E128-99 standards.