dNTP mix
Katalogové číslo 733-0548
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 mL
- Description:dNTP mix, 20 mM
Specifications
About this item
A combined mixture of dATP, dCTP, dGTP and dTTP containing 5 mM of each dNTP. Applications include PCR, RT-PCR, DNA labelling, and site-directed mutagenesis.