Bioindicator, Sterikon® plus
1.10274.0002
: Merck
- Description:Sterikon® plus bioindicator
- Pk:100 Ampoul
Sterikon® plus bioindicators can be used to test the functionality of autoclaves (15 minutes at 121 °C). They can also be used to check any sort of autoclave load for the success of sterilisation. For example, pharmaceuticals, particularly products in ampoules, canned food and more.
- Easy interpretation due to colour change
- Safe, because spores of non-pathogenic organisms (Geobacillus stearothermophilus) are used
Acc. to the recommendation of USP and EP for the use of bioindicators for checking the autoclaving of pharmaceutical products.
: According to the recommendation of USP and EP for the use of bioindicators for checking the autoclaving of pharmaceutical products.