Conductivity standards, Certipur® Supelco®
Supplier: Merck
Certificates
About this item
These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.
- Traceable to PTB and NIST
- Tested against primary reference standards
Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberCertipur
Product Literature
ProductPage_101203 ProductPage_101254 ProductPage_101255 ProductPage_101255 ProductPage_101553
Product specification
More
Frequently Bought Together