About this item
ROBU®® filter candles with integrated VitraPRO® filter, conical shape and narrow glass tube are ideal for precise, compact filtration. Designed for easy integration into laboratory setups, offering chemical resistance and reliable particle retention.
Integrated VitraPOR® porous glass
- Conical sealing surface
- Narrow glass outlet tube
- High chemical and thermal resistance
- Suitable for gas and liquid filtration
- Reusable and autoclavable
-
Uniform pore structure
Material : Borosilicate glass 3.3
ROBU filter candles with VitraPOR filter in conical shape with a narrow glass tube are engineered for use in confined or specialised filtration systems. The conical design allows a secure, leak-tight connection, while the narrow outlet tube enables targeted guidance of filtered media. Manufactured from chemically resistant borosilicate glass and equipped with a VitraPOR filter body, they deliver consistent filtration performance with high thermal and mechanical stability. Suitable for a wide range of applications in laboratories, analytics, and process technology.
In compliance with ISO 4796-80 and ASTM E128-99 standards.