ContactUsLinkComponent
VWR® Pipette Rack
VWR® Pipette Rack
Catalog # 612-3848
Supplier:  VWR Collection
undefined
VWR® Pipette Rack
Catalog # 612-3848
Supplier:  VWR Collection
Specifications
  • Colour:
    Blue
  • Description:
    Cargo manual pipette racks
  • W×D×H:
    95×127,5×301 mm
  • Pk:
    3
Specifications
About this item
Ideal for serological pipettes.
  • Click together horizontally with side connections
  • Securely lock in place with top connections
  • Magnets on both sides of units connect securely and safely to metal surface, such as filling cabinets, fume hoods, refrigerators and freezers
  • Angled shelves store 0,1 to 50 ml pipettes within arm's reach
  • Protective lid encloses the unit to prevent dust build up, and provides an additional shelf for extra space when connected horizontally