Order Entry
Czech Republic
ContactUsLinkComponent
 
Affinity chromatography media, Glutathione Sepharose™ 4 Fast Flow
 :  Cytiva
 
undefined
Affinity chromatography media, Glutathione Sepharose™ 4 Fast Flow
 :  Cytiva

 

  • pH range:
    3 - 12
  • Matrix:
    Highly cross-linked 4% agarose
  • Ligand:
    Glutathione and 10-carbon linker arm
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    >10 mg/ml
  • Flow rate:
    <450 cm/h

 

Frequently Bought Together