Printer and accessories for balances
The Sartorius printer YDP20-0CE has date, time statistics and transaction counter functions and is suitable for use in legal metrology. These capabilities when attached to balances with built in GLP functions, allow GLP compliant printouts including operator details and spaces for signatures.
Compatible with: Sartorius CP, CPA, LA, LP, LC, MC, ME, AC, PT, BP, TE, LE, SE, ED and MA balances.
SartoCollect: The standard data software for Sartorius instruments that facilitates data transfer to a PC with direct integration in MS Excel®, Word® and Access® or an ASCII file.
Other accessories are suitable for all ME, SE, LA, CP, CPA and TE models.
Frequently Bought Together