2-(2-Bromoethyl)-1,3-dioxolane stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18742-02-4
:
Formula:C₅H₉BrO₂
MDL Number:MFCD00003216
:181.03 g/mol
:M-Clarity™ quality level = MQ200
EINECS:242-551-1
Frequently Bought Together