1,2-Bis(phenylthio)ethane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:622-20-8
:
Formula:C₁₄H₁₄S₂
MDL Number:MFCD00014085
:246.4 g/mol
Melting Pt:68…70 °C
Storage Temperature:Ambient
EINECS:210-723-5
Related Information
Frequently Bought Together