



Potassium-tert-butoxide 1 M in THF, AcroSeal®, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:865-47-4
:
Formula:C₄H₉KO
MDL Number:MFCD00012162
:112.21 g/mol
Density:0.902 g/cm³
Flash Pt:–19 °C
Storage Temperature:Ambient
EINECS:212-740-3
UN:2920
ADR:8,I
:GHS05
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to yellow to light orange Solution (May develop some turbidity or precipitate)
- Strength:0.95 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together