Organic reference standard, Vanillic acid 10 µg/ml in methanol
Catalog # PROONE9341
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:121-34-6
- Description:Vanillic acid 10 µg/ml in methanol
- Formula:C₈H₈O₄
- Description:Vanillic acid