Pyruvaldehyde (methylglyoxal) 40% in aqueous solution, Sigma-Aldrich®
: Merck
:78-98-8
:
:C₃H₄O₂
:MFCD00006960
:M-Clarity™ quality level = MQ300
:72.06 g/mol
:201-164-8
Frequently Bought Together
Water, TECHNICAL, demineralised
: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
: VWR Chemicals
n-Hexane, EMPLURA®, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
: VWR Chemicals
Dichloromethane ≥99.5% stabilised ACS
: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals