Pyruvaldehyde (methylglyoxal) 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-98-8
:
Formula:C₃H₄O₂
MDL Number:MFCD00006960
:M-Clarity™ quality level = MQ200
:72.06 g/mol
EINECS:201-164-8
Frequently Bought Together