Glutaconic acid (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL49360-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:130.1 g/mol
EINECS:217-027-0
CAS Number:1724-02-3
Formula:C₅H₆O₄