2-(4-Chlorobutyl)-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIAL24045-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:150.6 g/mol
CAS Number:118336-86-0
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00043136