
2-(Bromomethyl)-1,3-dioxolane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4360-63-8
:
Formula:C₄H₇BrO₂
MDL Number:MFCD00003214
:167 g/mol
Boiling Pt:175 °C
Density:1.628
Flash Pt:62 °C (145 °F)
EINECS:224-443-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together