
2-(Bromomethyl)-1,3-dioxolane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4360-63-8
:
Formula:C₄H₇BrO₂
MDL Number:MFCD00003214
:167 g/mol
Boiling Pt:80…82 °C (27 mmHg)
Flash Pt:62 °C
EINECS:224-443-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4810 to 1.4840 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together