
2-(2-Bromoethyl)-1,3-dioxolane 96%
:18742-02-4
:
:C₅H₉BrO₂
:MFCD00003216
:181.03 g/mol
:68…70 °C (8 mmHg)
:1.54 g/cm³
:65 °C
:Freezer
:242-551-1
:6.1,III
:GHS07
- Appearance:Clear colorless to yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4780 to 1.4830 (20°C, 589 nm)
- GC:≥95 %
Frequently Bought Together