ContactUsLinkComponent
Boron standard solution, 1000 mg/L B in ammonium hydroxide solution (max. 1%) (from B(OH₃)), standard for ICP, Specpure®
Boron standard solution, 1000 mg/L B in ammonium hydroxide solution (max. 1%) (from B(OH₃)), standard for ICP, Specpure®
Certificates
Boron standard solution, 1000 mg/L B in ammonium hydroxide solution (max. 1%) (from B(OH₃)), standard for ICP, Specpure®
Certificates
Specifications
  • MDL No.:
    MFCD00134034