
Ammonium tetrachlorocuprate dihydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10060-13-6
:
Formula:H₈Cl₄CuN₂·2H₂O
MDL Number:MFCD00150384
:277.46 g/mol
Merck Index:14,514
Melting Pt:110 °C (dec.)
Density:1.993
EINECS:239-690-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together