2-Amino-4-hydroxy-6-methylpyrimidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3977-29-5
:
Formula:C₅H₇N₃O
MDL Number:MFCD00006095
:125.13 g/mol
:M-Clarity™ quality level = MQ100
EINECS:223-612-1
Frequently Bought Together