
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2564-83-2
:
Formula:C₉H₁₈NO
MDL Number:MFCD00009599
:156.25 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:219-888-8
UN:3263
:GHS05
Storage Temperature:Ambient
Frequently Bought Together