4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO), Sigma-Aldrich®
Catalog # SIAL390380-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:213.3 g/mol
:M-Clarity™ quality level = MQ200
EINECS:423-840-3
CAS Number:14691-89-5
Formula:C₁₁H₂₁N₂O₂
MDL Number:MFCD00043593