ContactUsLinkComponent
sec-Butyllithium 1.3 M in cyclohexane (92% )(v/v)/hexane (mixture of isomers) (8% (v/v)), AcroSeal®
Irritant
Health
Corrosive
Environment
Flammable
sec-Butyllithium 1.3 M in cyclohexane (92% )(v/v)/hexane (mixture of isomers) (8% (v/v)), AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Environment
Flammable
sec-Butyllithium 1.3 M in cyclohexane (92% )(v/v)/hexane (mixture of isomers) (8% (v/v)), AcroSeal®
Specification Test Results
  • Appearance:
    Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    10.5 to 12.5 %
  • UV:
    at 210 nm A: ≤0.07
  • Gradient analysis:
    pass test