2-Hydroxyethyl salicylate, Sigma-Aldrich®
Catalog # SIAL03830-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:182.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-737-2
CAS Number:87-28-5
Formula:C₉H₁₀O₄
MDL Number:MFCD00002862