L(+)-Ornithine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3184-13-2
:
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00064562
:168.62 g/mol
Melting Pt:245 °C (dec.)
Storage Temperature:Ambient
EINECS:221-678-6
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- Solubility:(5 % in 1 N HCl) Clear colorless
- Specific optical rotation:+21° to +24° (20°C, 589 nm) (c=5, 1 N HCl)
Specification Test Results
Frequently Bought Together