Beryllium standard solution, 1,000 mg/L Be in 1 M hydrochloric acid standard for AAS
Catalog # REGEAABEH
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HCl
- Pack type:Glass bottle
- Pk:500 mL
- Element:Beryllium
- Symbol:Be