Order Entry
Czech Republic
ContactUsLinkComponent
Barium standard solution, 10 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Barium standard solution, 10 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Catalog # 45247.AE
CAS Number:  
Barium standard solution, 10 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Catalog # 45247.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item

Traceable to NIST

Certifications: Traceable to NIST