Order Entry
Czech Republic
ContactUsLinkComponent
Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Flammable
Irritant
Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Catalog # 41908.04
CAS Number:  
Flammable
Irritant
Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Catalog # 41908.04
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g

Specifications

About this item

CAS Number:2172-02-3
 :
Formula:C₁₆H₃₆HO₄
MDL Number:MFCD00070458
 :470.94 g/mol
Boiling Pt:90 °C (5 mmHg)
Melting Pt:2 °C
Density:1.15
Flash Pt:28 °C (82 °F)
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
Formula:C₁₆H₃₆HfO₄