4,6-Dichloro-5-nitropyrimidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4316-93-2
:
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00006107
:193.98 g/mol
:M-Clarity™ quality level = MQ100
EINECS:224-340-6
Frequently Bought Together