
2-Chloro-4-methylpyrimidine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13036-57-2
:
Formula:C₅H₅ClN₂
MDL Number:MFCD00054434
:128.56 g/mol
Melting Pt:45…48 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together