Methyl oxalyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5781-53-3
:
Formula:C₃H₃ClO₃
MDL Number:MFCD00000705
:122.51 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:227-307-4
UN:2920
Frequently Bought Together