Order Entry
Czech Republic
ContactUsLinkComponent
Tin(II) chloride, anhydrous ≥98%
Corrosive
Health
Irritant
Tin(II) chloride, anhydrous ≥98%
Catalog # ABCRAB122425.0500
Supplier:  abcr
CAS Number:  
Corrosive
Health
Irritant
Tin(II) chloride, anhydrous ≥98%
Catalog # ABCRAB122425.0500
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :189.62 g/mol
Merck Index:13,08861
Boiling Pt:623 °C
Melting Pt:246.8 °C
Density:3.95 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,II
 :GHS05
 :GHS08
 :GHS07
CAS Number:7772-99-8
Formula:SnCl₂
MDL Number:MFCD00011241