
Divinylbenzene (mixture of isomers) 55% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1321-74-0
:
Formula:C₁₀H₁₀
MDL Number:MFCD01778656
:130.19 g/mol
Density:0.912
Flash Pt:62 °C (143 °F)
:Stabilised with 4-tert-butylpyrocatechol (TBC) 1000 ppm
EINECS:215-325-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together